Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Vial Seals
Easy to use, sterile flip seal
Manufacturer:  DWK Life Sciences W986011S
 View more versions of this product
Catalog No. 06-451-249
Add to cart
Description
Description
  • Autoclavable
  • Tamper-evident
Tamper-evident
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Flip Seal
220
Red
Aluminum
20mm
5mL or 10mL Serum Vial
Wheaton CompletePAK
20 mm
Sterile