Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Vial Seals
Easy to use, sterile flip seal
$329.75 - $349.68
Specifications
Sterility Sterile
Product Line Wheaton CompletePAK
Certifications/Compliance USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
View More Specs
Products 4
Catalog Number Mfr. No. Color Diameter (Metric) Outer For Use With Price Quantity & Availability  
Catalog Number Mfr. No. Color Diameter (Metric) Outer For Use With Price Quantity & Availability  
06-451-247 View Documents DWK Life Sciences
W986009S
Red 13 mm 2mL Serum Vial Case of 230 for $329.75
Add to cart
06-451-248 View Documents DWK Life Sciences
W986010S
Blue 13 mm 2mL Serum Vial Case of 230 for $329.75
Add to cart
06-451-249 View Documents DWK Life Sciences
W986011S
Red 20 mm 5mL or 10mL Serum Vial Case of 220 for $349.68
Add to cart
06-451-250 View Documents DWK Life Sciences
W986012S
Blue 20 mm 5mL or 10mL Serum Vial Case of 220 for $349.68
Add to cart
Description
Description
  • Autoclavable
  • Tamper-evident
Tamper-evident
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
Sterile
Wheaton CompletePAK
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified