Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Air-Tite™ Sterile Hypodermic Needles

Air-Tite™ Sterile Hypodermic Needles are high quality, yet economical for laboratory and veterinary use only
Manufacturer: Air-Tite™ N22112
Description
- Disposable Needles
- Sterile
- Polypropylene Hub
- Stainless Steel
- Single-use
- Latex Free
- ISO
- Lab/Vet
Specifications
No | |
Black | |
Laboratory / Veterinary | |
1.5 in. | |
100/Box | |
Leur | |
Luer Lock | |
Yes | |
Sterile |
ISO 9001 Certified | |
22 G | |
Yes | |
Stainless Steel | |
Individually Wrapped | |
Tri-Bevel | |
No | |
Polypropylene | |
Yes |
Safety and Handling
ShelfLife : 5 years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Not for human use - laboratory and veterinary use only