Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Vial Stoppers
Provide airtight covers
$316.42 - $541.22
Specifications
Sterility Sterile
Certifications/Compliance USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
View More Specs
Products 6
Catalog Number Mfr. No. Closure Size Closure Type Price Quantity & Availability  
Catalog Number Mfr. No. Closure Size Closure Type Price Quantity & Availability  
06-451-241 View Documents DWK Life Sciences
W986003S
13mm Omni Flex 3G Straight Plug Stopper Case of 230 for $467.57
Add to cart
06-451-244 View Documents DWK Life Sciences
W986006S
20mm Omni Flex 3G Straight Plug Stopper Case of 220 for $482.26
Add to cart
06-451-242 View Documents DWK Life Sciences
W986004S
13mm Uncoated Straight Plug Stopper Case of 230 for $371.44
Add to cart
06-451-245 View Documents DWK Life Sciences
W986007S
20mm Uncoated Straight Plug Stopper Case of 220 for $316.42
Add to cart
06-451-243 View Documents DWK Life Sciences
W986005S
13mm Igloo Lyo Stopper Case of 230 for $499.39
Add to cart
06-451-246 View Documents DWK Life Sciences
W986008S
20mm Igloo Lyo Stopper Case of 220 for $541.22
Add to cart
Description
Description
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
Sterile
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified