Learn More
BRAND™ Sterile PD-Tip™ II Precision Dispenser Tips for Repeating Pipettes PROMO
Fits all instruments using standard repeating tips.
Specifications
Certifications/Compliance Certificate of Performance/Purity Certificate
Clean Claims Free from endotoxin, DNA, Rnase, ATP
Autoclavable Not Autoclavable
Color Natural
Compatibility All standard repeating pipettes
View More Specs
Products 10
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
14-380-987PM
View Documents
BRAND™
705730IP
0.1 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-988PM
View Documents
BRAND™
705732IP
0.5 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-989PM
View Documents
BRAND™
705734IP
1 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-990PM
View Documents
BRAND™
705736IP
1.25 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-991PM
View Documents
BRAND™
705738IP
2.5 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-992PM
View Documents
BRAND™
705740IP
5 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-993PM
View Documents
BRAND™
705742IP
10 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-994PM
View Documents
BRAND™
705744IP
12.5 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-995PM
View Documents
BRAND™
705746IP
25 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-380-996PM
View Documents
BRAND™
705748IP
50 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Automatic size recognition by BRAND™ HandyStep™ S, HandyStep™ Touch S and HandyStep™ Electronic pipettes
  • BIO-CERT™ sterile
  • BIO-CERT™ tips are free of DNA, RNase, endotoxin and ATP
  • 25 mL and 50 mL sizes include one adapter
  • Adapters are autoclavable at 121°C for 20 minutes
Specifications
Specifications
Certificate of Performance/Purity Certificate
Not Autoclavable
All standard repeating pipettes
Individually Wrapped
Positive Displacement
HandyStep™ Touch and HandyStep Touch S
Polypropylene cylinder; Polyethylene plunger
Beta
Free from endotoxin, DNA, Rnase, ATP
Natural
Non-filtered
Sterile
Yes
Yes
PD-Tip II