Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Manufacturer: Thermo Scientific™ 91826105

 View more versions of this product

Catalog No. 21-402-636

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Color Clear
    For Use With (Equipment) Beckman Liquid Handling Systems
    Volume (Metric) 20μL
    Product Line Beckman
    Filtered No
    Sterility Sterile
    Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

    Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.  Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Beckman Biomek FX, Biomek NX, Biomek 3000 and Multimek AP96.

    Features:

    • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
    • Lot-tested during production on actual workstation for which they are created
    • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
    • Low percentage coefficient of variation (%CV)

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit