Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase performance through the superior tip straightness and low %CV with Thermo Scientific™ Sterile Pipette Tips for  Beckman™ Liquid Handling Systems.
Manufacturer:  Thermo Scientific™ 938-261
 View more versions of this product
Catalog No. 14-754-461
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Increase pipetting performance with Thermo Scientific? Sterile Pipette Tips for Beckman? Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)
Description & Specifications
Specifications
250μL
Sterile
Beckman Liquid Handling Systems
No
384 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
ART