Learn More
AZER Scientific Sterile Robotic Conductive Tips (Hamilton Compatible)
Designed to be compatible with Hamilton Microlab Star™, Nimbus™ Vantage™ pipetting instruments.
$658.17 - $1142.27
Specifications
Color Black
Material PP (Polypropylene)
Sterility Sterile
Autoclavable No
View More Specs
Products 6
Catalog Number Mfr. No. Filtered Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Filtered Volume (Metric) Price Quantity & Availability  
22-026-004 View Documents AZER Scientific
ES490101004
Non-Filtered 50μL Case of 7680 for $1,031.03
Add to cart
22-026-005 View Documents AZER Scientific
ES490101104
Filtered 50μL Case of 7680 for $1,142.27
Add to cart
22-026-009 View Documents AZER Scientific
ES490081004
Non-Filtered 300μL Case of 7680 for $1,031.03
Add to cart
22-026-010 View Documents AZER Scientific
ES490081104
Filtered 300μL Case of 7680 for $1,142.27
Add to cart
22-026-013 View Documents AZER Scientific
ES490091004
Non-Filtered 1000μL Case of 4800 for $658.17
Add to cart
22-026-014 View Documents AZER Scientific
ES490091104
Filtered 1000μL Case of 4800 for $725.12
Add to cart
Description
Description
Conductivity enables the system to recognize the filling height to assure minimal immersion of the tip into the liquid.
  • Manufactured under cleanroom conditions
  • Each lot tested according to strict quality management system to ensure reproducible results for every application
  • High quality, tested by independent labs to be free of DNase, RNase, ATP and pyrogens
  • Conductive tips allow pipettor to sense liquid levels
  • Made from pure Polypropylene
Specifications
Specifications
Black
PP (Polypropylene)
Sterile
No