Learn More
Research Products International CorpSupplier Diversity Partner Sterile Syringe Filters, Disposable, PES Membrane, .22um Pore Size, 13mm Diameter, 75 per Pack DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 256130
Syringe filters are ideal for filtering culture media and organic solvents in a wide range of applications. All items are tested for filter efficiency and housing integrity. Filter housing is manufactured and tested to withstand pressures of up to 75 PSI (5.1 Bar). Mainly usedfor pre-filtrationandpurification to sterilize and make samples of solution particle-free andsterile. Designed for low hold-up to minimize sample loss. Lot numbers are printed on sterile packs for easy traceability. Sterile filters are packaged in a convenient, peel-open package. All products come certified non-pyrogenic and Gamma irradiated. RNase and DNase free.
Catalog No. 50-476-572
Due to product restrictions, please to purchase or view availability for this product.