Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Type 1 Clear Serum Vials Available on GSA/VA Contract for Federal Government customers only.
Made with Type I Borosilicate glass
Manufacturer:  DWK Life Sciences W986000S
 View more versions of this product
Catalog No. 06-451-238
Add to cart
Description
Description
  • Type I glass serum vials
  • Sterile
  • Particulate cleaned, dehydrogenated, and sterilized certified to USP <788>, <85> & <71>
  • Container Closure Integrity (CCI) certified
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Vial
Borosilicate Glass (Type I)
2 mL
Wheaton CompletePAK
13mm
Clear
Sterile
210