Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Type 1 Clear Serum Vials
Made with Type I Borosilicate glass
Manufacturer:  DWK Life Sciences W986001S
 View more versions of this product
Catalog No. 06-451-239
Add to cart
Description
Description
  • Type I glass serum vials
  • Sterile
  • Particulate cleaned, dehydrogenated, and sterilized certified to USP <788>, <85> & <71>
  • Container Closure Integrity (CCI) certified
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
Clear
Sterile
210
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Borosilicate Glass (Type 1)
5 mL
Wheaton™ CompletePAK
20 mm