Learn More
DWK Life Sciences Wheaton™ CompletePAK Sterile Type 1 Clear Serum Vials Available on GSA/VA Contract for Federal Government customers only.
Made with Type I Borosilicate glass
$425.63 - $592.61
Specifications
Color Clear
Material Borosilicate Glass (Type I)
Sterility Sterile
Product Line Wheaton CompletePAK
Type Vial
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Closure Size Price Quantity & Availability  
06-451-238 Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
W986000S
2 mL 13mm Case of 210 for $425.63
Add to cart
06-451-239 View Documents DWK Life Sciences
W986001S
5 mL 20mm Case of 200 for $459.18
Add to cart
06-451-240 Available on GSA/VA Contract for Federal Government customers only. View Documents DWK Life Sciences
W986002S
10 mL 20mm Case of 158 for $592.61
Add to cart
Description
Description
  • Type I glass serum vials
  • Sterile
  • Particulate cleaned, dehydrogenated, and sterilized certified to USP <788>, <85> & <71>
  • Container Closure Integrity (CCI) certified
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Clear
Sterile
Vial
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Borosilicate Glass (Type I)
Wheaton CompletePAK
210