Learn More
Globe Scientific Sterile Universal Filter Tips
Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems.
$248.50 - $386.00
Specifications
Clean Claims Rnase, Dnase, and Pyrogen Free
Filtered Filtered
Format Rack
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Length (Metric) Price Quantity & Availability  
22-171-777
View Documents
Globe Scientific
150900
0.1 to 10 μL 32 mm
Case of 1920 for $248.50
Add to cart
22-171-779
View Documents
Globe Scientific
150910
1 to 20 μL 50 mm
Case of 1920 for $276.00
Add to cart
22-171-780
View Documents
Globe Scientific
150915
1 to 100 μL 50 mm
Case of 1920 for $276.00
Add to cart
22-171-781
View Documents
Globe Scientific
150920
1 to 200 μL 59 mm
Case of 1920 for $276.00
Add to cart
22-171-782
View Documents
Globe Scientific
150925
1 to 300 μL 59 mm
Case of 1920 for $276.00
Add to cart
22-171-783
View Documents
Globe Scientific
150935
1 to 1000 μL 86 mm
Case of 1920 for $386.00
Add to cart
Description
Description
  • Manufactured from clear polypropylene (PP)
  • Polypropylene (PP) filters protect samples and pipettes from contamination
  • Dnase-free, Rnase-free and non-pyrogenic
  • Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
  • Sterilized by gamma irradiation SAL10-6 (ISO11137)
  • Racks are printed with lot number for quality traceability
Specifications
Specifications
Rnase, Dnase, and Pyrogen Free
Filtered
Rack