Learn More
Globe Scientific Sterile Universal Filter Tips, Extended Length
Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems.
Supplier: Globe Scientific 150903
Description
- Manufactured from clear polypropylene (PP)
- Polypropylene (PP) filters protect samples and pipettes from contamination
- Dnase-free, Rnase-free and non-pyrogenic
- Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
- Sterilized by gamma irradiation SAL10-6 (ISO11137)
- Racks are printed with lot number for quality traceability
Specifications
0.1 to 10 μL | |
Sterile | |
Not Autoclavable | |
Rnase, Dnase, and Pyrogen Free | |
Rack | |
96 Tips/Rack, 10 Racks/Pk, 2 Boxes/Cs. | |
Yes |
Filtered | |
Natural | |
45 mm | |
Gamma Irradiation | |
Extended Length | |
Universal Fit |