Learn More
Globe Scientific Sterile Universal Filter Tips, Extended Length
Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems.
$248.50 - $474.50
Specifications
Clean Claims Rnase, Dnase, and Pyrogen Free
Filtered Filtered
Format Rack
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Length (Metric) Price Quantity & Availability  
22-171-778
View Documents
Globe Scientific
150903
0.1 to 10 μL 45 mm
Case of 1920 for $248.50
Add to cart
22-171-784
View Documents
Globe Scientific
150938
1 to 1000 μL 105 mm
Case of 1920 for $474.50
Add to cart
Description
Description
  • Manufactured from clear polypropylene (PP)
  • Polypropylene (PP) filters protect samples and pipettes from contamination
  • Dnase-free, Rnase-free and non-pyrogenic
  • Manufactured in a Class 100,000 cleanroom environment under ISO13485:2016 and ISO9001:2015 quality management systems
  • Sterilized by gamma irradiation SAL10-6 (ISO11137)
  • Racks are printed with lot number for quality traceability
Specifications
Specifications
Rnase, Dnase, and Pyrogen Free
Filtered
Rack