Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sterile Water for Injection, Meets USP Specifications, Spectrum™

Click to view available options
Quantity:
1 L
20 L
200 L
7732-18-5
7732-18-5
Grade Reagent
CAS Max % 1
Bacterial Endotoxins 0.25 EU/mL
Quantity 1 L

CAUTION: For further manufacturing, processing, repacking or use in laboratories or research. Not intended for direct human or veterinary parenteral administration. Read and understand the label and Safety Data Sheet (SDS) prior to use.