Learn More
Research Products International Corp
Sterile Cloning Discs, 3.2mm (1/8 Inch) Diameter, 100 per Package
Manufacturer: Research Products International Corp 248710
Convenient method for isolating a large number of clones. Sterile paper discs allow transfer of cells to a multi-well plate in a fraction of the time normally required and reduce possibility of contamination. Discs are sterilized by Gamma radiation. Supplied in two sterile tubes each containing fifty discs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.