Learn More
Research Products International CorpSupplier Diversity Partner Sterile Cloning Discs, 5.0mm (3/16 Inch) Diameter, 100 per Package  DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 248711
Convenient method for isolating a large number of clones. Sterile paper discs allow transfer of cells to a multi-well plate in a fraction of the time normally required and reduce possibility of contamination. Discs are sterilized by Gamma radiation. Supplied in two sterile tubes each containing fifty discs.
Catalog No. 50-550-299