Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Sterileware™ Flat Bottom Sampling Scoop

Bel-Art™ Sterileware™ Flat Bottom Sampling Scoop is used by pharmaceutical, food, cosmetic and chemical companies for general production or laboratory work.

Manufacturer:  Bel-Art™ F369110150

 View more versions of this product

Catalog No. 03-411-794


Add to cart

Description

Description

  • Flat bottom allows for use as a weigh boat
  • Approximate volumes can be measured by removing excess powder
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing
  • Shipped with a Certificate of Processing for Sterilization
Specifications

Specifications

High-impact Polystyrene
Bowl: 12.3 x 6.8 x 3.4 cm (4 7/8 x 2 3/4 x 1 3/8 in.)
Flat
5.1 oz.
100/Pk.
Sterile
Yes
9.5 cm
3 3/4 in.
150 mL
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit