Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
Manufacturer:  Thermo Scientific™ 511
 View more versions of this product
Catalog No. 51-1
Add to cart
Description
Description
Thermo Scientific™ Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
NOT AVAILABLE IN AMERICA
  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
Specifications
Specifications
Autoclave bags (HDPE)
200
32 in.
HDPE (High-density Polyethylene)
610mm
810 x 610mm
121°C
810mm
24 in.