Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
$53.60
Products 1
Catalog Number Mfr. No. Description Temperature Range (Metric) Dimensions (L x W) No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Description Temperature Range (Metric) Dimensions (L x W) No. per Case Price Quantity & Availability  
51-1
View Documents
Thermo Scientific™
511
Autoclave bags (HDPE) 121°C 810 x 610mm 200 Each for $53.60
Add to cart
Description
Description
Thermo Scientific™ Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
NOT AVAILABLE IN AMERICA
  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions