Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Steris™ Alcare Plus™ Antiseptic Handrub with Emollients

Fast-acting, foamed, alcohol-based handwash with added emollients that moisturize skin

Manufacturer:  Steris™ 639957

 View more versions of this product

Catalog No. 14-415-26



Add to cart

Description

Description

Ideal for use when soap and water are not available.

  • Contains 62% ethyl alcohol by volume
  • Provides broad spectrum activity against pathogenic microorganisms, including methicillin-resistant Staphylococcus aureus (MRSA) and vancomycin-resistant enterococci (VRE)
  • Spreads easily to facilitate even distribution over all skin surfaces for effective disinfection

Specifications

Specifications

FDA
Provides broad spectrum activity against pathogenic microorganisms, kills 99.99% of mrsa and vre in 5 sec.
5.61 oz.
Each
Ethyl Alcohol 62%
Alcare Plus™ Antiseptic Handrub
160mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit