Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Steris™ Cal Stat™ Plus Antiseptic Handrub

Quickly decontaminates visibly clean hands

$66.40

Specifications

Certifications/Compliance FDA
Quantity 1L
View More Specs

Requires: 14-415-27 requires use of Steris Dispensing System (14-415-21)


Products 3
Catalog Number Mfr. No. Quantity For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Quantity For Use With (Application) Price Quantity & Availability  
14-415-27
View Documents Promotion Details
Steris™
118187
1L For Alcohol-based waterless healthcare personnel handwash
Each for $66.40
Case of 12 EA for $631.50
Add to cart
 
14-415-28
Required Accessories View Documents Promotion Details
Steris™
1181RP
444mL For Alcohol-based waterless healthcare personnel handwash This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
14-415-29
View Documents Promotion Details
Steris™
118103
118mL Kill of a broad spectrum of transient microorganisms, for Alcohol-based waterless healthcare personnel handwash This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • 63% v/v isopropyl-alcohol base
  • Emollients moisturize skin
  • Does not inactivate chlorhexidine gluconate (CHG) residual on skin
  • Dermatologist-tested
  • Dye-free
  • Hypoallergenic

Specifications

Specifications

FDA
1L
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit