Learn More
Steris™ Coverage™ Spray TB Plus Ready-to-Use Disinfectant Cleaner
Hard surface quaternary-based disinfectant kills a broad range of bacteria, viruses, and fungi within three minutes
Manufacturer:  Steris™ 1629B4
 View more versions of this product
Catalog No. 14-415-12
Add to cart
Description
Description
Ideal for use in laboratories, hospitals, surgical centers, medical and dental offices, and other patient-care facilities.
  • Tuberculocidal and HIV-1 effective
  • Leaves no hazardous residue
  • Contains no dangerous phenols or chlorine
  • Compatible for use on inanimate surfaces including plastics, stainless steel, chrome, vinyl, acrylic, glass, Plexiglas™ resin, and baked-on painted surfaces
  • Uses a sprayer head on the 24 oz. (710mL) bottle that allows application without producing aerosolized airborne particles that may be aspirated
Compliance
Conforms to OSHA Guidelines (21CFR Part 1910, 1030).
Specifications
Specifications
24 oz.
OSHA
21% Isopropanol
Kills a broad range of Bacteria, Viruses and Fungi on inanimate surfaces including Plastics, Stainless Steel, Chrome, Vinyl, Acrylic, Glass, Plexiglas and baked-on painted surfaces
6.5 to 8.7
710 mL
Clear
Trigger spray bottle
Liquid