Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Steris Coverage Spray TB Plus Ready-to-Use Disinfectant Cleaner
Hard surface quaternary-based disinfectant kills a broad range of bacteria, viruses, and fungi within three minutes
Specifications
Certifications/Compliance | OSHA |
---|---|
Color | Clear |
For Use With (Application) | Kills a broad range of Bacteria, Viruses and Fungi on inanimate surfaces including Plastics, Stainless Steel, Chrome, Vinyl, Acrylic, Glass, Plexiglas and baked-on painted surfaces |
Format | Liquid |
pH Range | 6.5 to 8.7 |
Catalog Number | Mfr. No. | Capacity (Metric) | Capacity (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Capacity (English) | Price | Quantity & Availability | ||||
14-415-12
|
Steris
1629B4 |
710 mL | 24 oz. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
14-415-13
|
Steris
162908 |
3.8 L | 1 gal. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
Ideal for use in laboratories, hospitals, surgical centers, medical and dental offices, and other patient-care facilities.
- Tuberculocidal and HIV-1 effective
- Leaves no hazardous residue
- Contains no dangerous phenols or chlorine
- Compatible for use on inanimate surfaces including plastics, stainless steel, chrome, vinyl, acrylic, glass, Plexiglas resin, and baked-on painted surfaces
- Uses a sprayer head on the 24 oz. (710mL) bottle that allows application without producing aerosolized airborne particles that may be aspirated
Compliance
Conforms to OSHA Guidelines (21CFR Part 1910, 1030).
Specifications
OSHA | |
Kills a broad range of Bacteria, Viruses and Fungi on inanimate surfaces including Plastics, Stainless Steel, Chrome, Vinyl, Acrylic, Glass, Plexiglas and baked-on painted surfaces | |
6.5 to 8.7 |
Clear | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title