Learn More
Steris™ Germicidal Surface Wipes
Nonwoven disposable cloths containing a stable, low pH disinfectant/deodorant cleaner
Manufacturer:  Steris™ 1608WC
 View more versions of this product
Catalog No. 14-415-17
Add to cart
Description
Description
  • Dual chain quaternary/isopropyl alcohol for use on hard, nonporous surfaces
  • Provide broad spectrum biocidal activity, including Mycobacterium tuberculosis and HIV-1 virus
  • Ready-to-use wipe—no mixing required
Designed for use on hard, nonporous surfaces such as stainless steel, formica, tables, carts, baskets, counters, cabinets, and telephones.
Specifications
Specifications
OSHA
White
10 x 10 in.
Tub of 60 wipes