Learn More
Steris™ Germicidal Surface Wipes
Nonwoven disposable cloths containing a stable, low pH disinfectant/deodorant cleaner
Specifications
Certifications/Compliance OSHA
Color White
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
14-415-16
View Documents
Steris™
1608GS
6 x 6.75 in. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
14-415-17
View Documents
Steris™
1608WC
10 x 10 in. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Dual chain quaternary/isopropyl alcohol for use on hard, nonporous surfaces
  • Provide broad spectrum biocidal activity, including Mycobacterium tuberculosis and HIV-1 virus
  • Ready-to-use wipe—no mixing required
Designed for use on hard, nonporous surfaces such as stainless steel, formica, tables, carts, baskets, counters, cabinets, and telephones.
Specifications
Specifications
OSHA
White