Learn More
Steris™ Lotion Soft™ Skin Conditioner Skin Cream
Moisturizes skin without compromising the antimicrobial activity of products containing chlorhexidine gluconate (CHG)
Supplier: Steris™ 1029K6
Requires : 14-415-30 requires use of Steris Dispensing System (14-415-21)
This item is not returnable. View return policy
Description
Moisturizes skin without compromising the antimicrobial activity of products containing chlorhexidine gluconate (CHG)
- Does not contain petrolatum or mineral oil—suitable for use under gloves
- Contains emollients and other softening ingredients that make it ideal for use after antimicrobial handwashing
Specifications
115 mL |