Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Steritop™ Threaded Bottle Top Filters
Click to view available options
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
Material (Membrane):
PES
PVDF
Description
The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.
- Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
- Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
- Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
- Sterilization of Aqueous Solutions (150 mL to 1 L)
- Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
Order Info
Note: Image above shows Bottle Top Filter with receiver flask (sold separately).

Specifications
Specifications
Color | Clear with blue adapter |
Disposable | Single-use |
Material | Polystyrene |
Material (Membrane) | PES |
Pore Size | 0.22 μm |
Sterility | Sterile |
For Use With (Application) | Sterile filtration |
Outlet Fitting | 45 mm |
Type | Bottle Top Filter |
Volume (Metric) | 150 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction