Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

STET Buffer, 1X Solution (pH 8.0), Fisher BioReagents

Specifications

Chemical Name or Material STET Buffer, 1X Solution
Name Note pH 8.0
CAS 77-86-1, 60-00-4, 7647-01-0, 7647-14-5, 9002-93-1
View More Specs

Products 2
Catalog Number Quantity Packaging Price Quantity & Availability  
Catalog Number Quantity Packaging Price Quantity & Availability  
BP24801
View Documents
1L Poly Bottle This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BP2480100
View Documents
100mL Poly Bottle This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

STET buffer is used in biomolecular procedures.
Specifications

Specifications

STET Buffer, 1X Solution
pH 8.0
77-86-1, 60-00-4, 7647-01-0, 7647-14-5, 9002-93-1
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit