Learn More
Drg International IncSupplier Diversity Partner Stfr Elisa Ruo DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA5925R
A sandwich enzyme immunoassay for the quantitative measurement of human soluble transferrin receptor. This product is for Research Use Only. In the Human sTfR ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with monoclonal anti-human sTfR antibody.
Catalog No. 50-194-7630