Learn More
STH Mouse anti-Human, Clone: OTI7A12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810382
Catalog No. 50-167-0046
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
STH is involved in positive regulation of mRNA splicing, via spliceosome.
Specifications
Specifications
STH
Monoclonal
Unconjugated
STH
Lyophilized
STH
Full length human recombit protein of human STH produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI7A12
PBS with 8% trehalose and no preservative; pH 7.3
Q8IWL8
MAPTIT
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
246744