Learn More
STIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152181AP
Catalog No. 50-173-0074
Add to cart
Description
Description
STIP1 is an adaptor protein that coordinates the functions of HSP70 and HSP90 in protein folding. It is thought to assist in the transfer of proteins from HSP70 to HSP90 by binding both HSP90 and substrate-bound HSP70. STIP1 also stimulates the ATPase activity of HSP70 and inhibits the ATPase activity of HSP90, suggesting that it regulates both the conformations and ATPase cycles of these chaperones (Song and Masison, 2005 ).
Specifications
Specifications
STIP1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35814, P31948, Q60864
HOP, Hsc70/Hsp90 organizing protein, IEF SSP 3521, P60, STI1, STI1L, STIP1
Rabbit
IgG
150 μL
-20°C
Primary
10963, 192277, 20867
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
STIP1
Liquid
Stip1
STIP1 Fusion Protein Ag7388
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat