Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Stir Light 7 x 7in. Stirrer

Mix reagents, stir bacteria cultures and perform other lab applications more effectively with these 7 x 7 ft. stirrers, which illuminate hard-to-see titration end points.

Manufacturer:  Thermo Scientific™ SL194325

Catalog No. 11-474-526

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mix reagents, stir bacteria cultures and perform other lab applications more effectively with the Thermo Scientific™ Stir Light 7 x 7 in. stirrer, which illuminates hard-to-see titration end points.

Key Features:
  • Lighted top allows easy viewing of samples while stirring
  • Light and stirring functions can be used independently
  • Variable stirring speeds from 250 to 2500rpm
  • Large 7 x 7in. acrylic top plate surface accommodates 4L flasks
  • Cool fluorescent bulbs minimize heat output
  • Features built-in indicator light

Includes: TFE-coated stir bar

Specifications

Specifications

37w
2.7kg
120V
60Hz
120V 60Hz
6 lb.
250 to 2500RPM
Stir Light 7 x 7 in. Stirrer
cCSAus
Stirrers
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit