Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Angled, Smooth, Stainless Steel. Sklar's Stitch Scissors are used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 153150
Sklar Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Angled, Smooth, Stainless Steel. Sklar's Stitch Scissors are used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Stitch scissors should not be used for cutting tissue. This product is angled with smooth, blunt/blunt tips and a length of 4-1/2 inches.
Catalog No. 50-143-7371