Learn More
Sklar InstrumentsSupplier Diversity Partner Sklarlite Spencer (Littauer) Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Scissors. Straight, Smooth, Extra Light (SklarLite), Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 231189
Sklarlite Spencer (Littauer) Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Scissors. Straight, Smooth, Extra Light (SklarLite), Stainless Steel. Sklarlite Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. Sklarlite instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This product is straight with smooth, blunt/blunt tips and a length of 4-1/2 inches.
Catalog No. 50-143-7947