Learn More
STK11, Rabbit anti-Human, Clone: 27D10, Abnova™
Rabbit Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB16197
Catalog No. 89-998-971
Add to cart
Description
Description
Specifications
Specifications
STK11
27D10
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Immunoprecipitation (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
serine/threonine kinase 11
LKB1/PJS
Rabbit
IgG
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
In 10 mM HEPES, 150 mM NaCl (50% glycerol, 100 ug/mL BSA, <0.02% sodium azide).
Liquid
STK11
A synthetic peptide (conjugated with KLH) corresponding to human STK11.
50 μL
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
6794