Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STK16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583089
Description
Immunogen sequence: CSRGTVIIDN KRYLFIQKLG EGGFSYVDLV EGLHDGHFYA LKRILCHEQQ DREEAQREAD MHRLFNHPNI LRLVAYCLRE RGAKHEAWLL LPFFKRGTLW NEIERLKDKG NFLTEDQILW LLLGICRGLE AIHAKGY.
STK16 (PKL12) is a serine/threonine kinase that functions in signal transduction pathways and transcriptional regulation. Protein kinase that act on both serine and threonine residues. STK16 may regulate transforming growth factor-beta (Tgfb) and vascular endothelial growth factor transcription.
Specifications
STK16 | |
Polyclonal | |
Unconjugated | |
STK16 | |
Edpk; Embryo-derived protein kinase; F52; hPSK; kinase related to cerevisiae and thaliana; Krct; MPSK; Mpsk1; Myristoylated and palmitoylated serine/threonine-protein kinase; PKL12; protein kinase expressed in day 12 fetal liver; Protein kinase Krct; Protein kinase PKL12; serine/threonine kinase 16; serine/threonine-protein kinase 16; Stk16; TGF-beta-stimulated factor 1; Tsf1; TSF-1; tyrosine-protein kinase STK16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8576 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75716 | |
STK16 | |
Recombinant protein corresponding to Human STK16. Recombinant protein control fragment (Product #RP-91412). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction