Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STK36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556326
Description
Immunogen sequence: SLLRHSQESN SLQQQSWYGT FLQDLMAVIQ AYFACTFNLE RSQTSDSLQV FQEAANLFLD LLGKLLAQPD DSEQTLRRDS LMCFTVLCE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
Serine/threonine protein kinase required for postnatal development, possibly by regulating the homeostasis of cerebral spinal fluid or ciliary function. Controls the activity of the transcriptional regulators GLI1, GLI2 and GLI3 by opposing the effect of SUFU and promoting their nuclear localization. GLI2 requires an additional function of STK36 to become transcriptionally active, but the enzyme does not need to possess an active kinase catalytic site for this to occur.
Specifications
STK36 | |
Polyclonal | |
Unconjugated | |
STK36 | |
1700112N14Rik; B930045J24; FU; Fused; Fused homolog; Kiaa1278; mKIAA1278; serine/threonine kinase 36; serine/threonine kinase 36 (fused homolog, Drosophila); serine/threonine-protein kinase 36; STK36; testicular tissue protein Li 188 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRP7 | |
STK36 | |
Recombinant protein corresponding to Human STK36. Recombinant protein control fragment (Product #RP-95074). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction