Learn More
STK38L Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI4D8, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA526073
Catalog No. PIMA526073
Add to cart
Description
Description
STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal. Northern blot analysis revealed expression of a 3.9-kb transcript in all tissues tested, with the possible exception of adult brain. Highest expression was in peripheral blood leukocytes. Immunoblot and immunofluorescence microscopy demonstrated predomitly nuclear expression of a 55-kD protein.
Specifications
Specifications
STK38L
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y2H1, Q7TSE6
NDR2, NDR2 protein kinase, nuclear Dbf2-related 2, nuclear Dbf2-related kinase 2, serine/threonine-protein kinase 38-like
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23012, 232533, 477663, 691337, 711047
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4D8
Unconjugated
STK38L
Liquid
STK38L
Human recombit protein fragment corresponding to amino acids 1-281 of STK38L produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat