Learn More
Aviva Systems Biology Corp STK39 Antibody - C-terminal region (ARP48745_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STK39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human STK39. Alternate names: DCHT, DKFZp686K05124, PASK, SPAKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59912