Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ STK4 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796475PLATES

 View more versions of this product

Catalog No. NB010334


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human STK4.
Specifications

Specifications

HRP
Antibody Pairs
93.75 pg/mL
6789
RUO
STK4
Apoptosis, Phospho Specific
Human
STK4
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 1.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 6000 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human STK4.
Sandwich ELISA
  • Detection Antibody: 0.1 mg/mL of mouse anti-Human STK4 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of mouse anti-Human STK4 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 200 ng of recombinant Human STK4.
  • Storage is content dependent.
    Mouse
    5 Plates
    93.75-6000 pg/mL
    EC 2.7.11, EC 2.7.11.1, kinase responsive to stress 2, KRS2KRS2)), Mammalian STE20-like protein kinase 1, mammalian sterile 20-like 1, MST-1, MST1DKFZp686A2068, serine/threonine kinase 4, serine/threonine-protein kinase 4, Serine/threonine-protein kinase Krs-2, STE20-like kinase MST1
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit