Learn More
MEDPLUS SERVICES USA STL STERI-DRAPE NO1089
Supplier:  MEDPLUS SERVICES USA MMM 1089
Encompass_Preferred
3M Steri-Drape Utility Sheet, Utility Sheet; 3M Biocade material, Overall Size: 22 inch x 25 inch (57cm x 66cm), used to help create a sterile field
Catalog No. NC9430913
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.