Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STMN2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521768

Catalog No. PIPA521768


Only null left
Add to Cart

Description

Description

Recommended positive controls: A549. Predicted reactivity: Mouse (100%), Rat (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

STMN2, also known as SCG-10, is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin. STMN2 may play a role in neuronal differentiation, and in modulating membrane interaction with the cytoskeleton during neurite outgrowth. Reductions in the expression of this gene have been associated with Down's syndrome and Alzheimer's disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STMN2
Polyclonal
Unconjugated
Stmn2
AI159727; hypothetical protein LOC534991; neuronal growth-associated protein (silencer element); neuron-specific growth-associated protein; Protein SCG10; SCG10; Scgn10; SGC10; stathmin 2; stathmin-2; stathmin-like 2; Stmb2; Stmn2; superior cervical ganglia, neural specific 10; superior cervical ganglion-10 protein; superiorcervical ganglia, neural specific 10
Rabbit
Antigen affinity chromatography
RUO
11075
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.75 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q93045
Stmn2
Synthetic peptide corresponding to a region within amino acids 2 and 41 of STMN2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.