Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STMN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 120271AP150UL

Description
RB3 (Stathmin-4), encoded by STMN4 gene, is a neuron-specific homologue of the SCG10/stathmin family proteins, possessing a unique N-terminal membrane-associated domain and the stathmin-like domain at the C terminus, which promotes microtubule (MT) catastrophe and/or tubulin sequestering. Stathmin is a ubiquitous cytosolic phosphoprotein, preferentially expressed in the nervous system, and described as a relay integrating diverse intracellular signaling pathways. RB3 is consistently up-regulated in differentiated neuroblastoma cells, and stable expression of transfected RB3 could induce differentiation, suggesting a role of RB3 during neuroblastoma differentiation. Moreover, increased RB3 is a unique response protein in the stathmin-related proteins following optical nerve axotomy and RB3 may be a critical target for neurodegenerative diseases.Specifications
STMN4 | |
Polyclonal | |
Unconjugated | |
STMN4 | |
RB3, Stathmin 4, stathmin like 4, Stathmin like protein B3, STMN4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56471, 79423, 81551 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63042, P63043, Q9H169 | |
Stmn4 | |
STMN4 Fusion Protein Ag2657 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction