Learn More
Stockwell Scientific Round-Bottom Test Tube
Ideal for routine laboratory analysis and procedures
Manufacturer:  Stockwell Scientific 8562L
 View more versions of this product
Catalog No. 03-342-22E
  Check Availability
Add to cart
Description
Description
  • Virtually unbreakable compared to similar glass tubes
  • Polypropylene (PP)
  • Chemically resistant
  • Autoclavable
  • 5mL capacity
  • 12 x 75mm
Specifications
Specifications
Lavender
5mL
75mm
Test Tube
12mm
5mL
Polypropylene (PP)
Round Bottom