Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoddard Solvent, Fisher Chemical™

Click to view available options
Quantity:
4L
200L
8052-41-3
MFCD00132767
8052-41-3
MFCD00132767

A petroleum distillate used for dry cleaning

Stoddard solvent, also known as mineral spirits, white spirit, petrolelum naptha, or paint thinner is a petroleum distillate used as a common solvent in paints, varnishes and asphalt, and in degreasing and dry cleaning. It is a mixture of aliphatic and alicyclic C7 to C12 hydrocarbons.
Acidity Pass Test
Boiling Range 90 to 100% less than 190°C
Physical Form Liquid
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 0.37 - 0.39 kPa at 20°C
Boiling Point 149°C
Color Colorless to Yellow
Quantity 4L
Chemical Name or Material Stoddard Solvent

WARNING!

Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:2
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid