Learn More
Stoko™ Esteso™ Classic Light-Duty Hand Cleanser
Gentle enough for everyday use
Manufacturer:  Stockhausen Inc. 8 83309 06
 View more versions of this product
Catalog No. 19-819-809
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Estesol™ Classic Hand Cleanser
67.6 oz., 2000mL
Clear, Light Green
General purpose restrooms and light manufacturing
67.62 oz.
Bottle, one pump
5 to 6
Green Seal Certified™
Water, Sodium Laureth Sulfate, Cocamidopropyl BetainePEG-6 Caprylic/Capric Glycerides, C12-C13 Alkyl Lactate, Decyl Glucoside, fragrance, 2-Bromo-2-Nitropropane-1,3-Diol, Citric Acid, blue 1, yellow 5
Pleasant
2000mL
9 Case
Safety and Handling
ShelfLife : 30 Month(s)