Stoko™ Esteso™ Classic Light-Duty Hand Cleanser

Gentle enough for everyday use

Manufacturer: Stockhausen Inc. 8 83311 06

 View more versions of this product

Catalog No. 19-819-810

  Check Availability
Add to Cart

    Specifications

    Size (Metric) 2000mL
    Packaging Bottle, soft
    Color Clear, Light Green
    For Use With (Application) General purpose restrooms and light manufacturing
    Fragrance Pleasant
    Shelf Life 30 Month(s)
    pH Range 5 to 6
    Size (English) 67.62 oz.
    Quantity 6 Case
    Certifications/Compliance Green Seal Certified™
    Composition Water, Sodium Laureth Sulfate, Cocamidopropyl BetainePEG-6 Caprylic/Capric Glycerides, C12-C13 Alkyl Lactate, Decyl Glucoside, fragrance, 2-Bromo-2-Nitropropane-1,3-Diol, Citric Acid, blue 1, yellow 5
    Product Type Estesol™ Classic Hand Cleanser

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit