Stoko™ Instant Hand Sanitizer

Great for use where water is unavailable

Manufacturer: Stockhausen Inc. 1 00888 06

 View more versions of this product

Catalog No. 19-141-650

  Check Availability
Add to Cart

    Specifications

    Size (English) 67.62 oz.
    Size (Metric) 2000mL
    Packaging Bottle, soft
    Quantity 6 Case
    Color Clear
    Composition Ethanol 62%, Deionized Water, Isopropanol, Hydrogenated Starch, THP Ethylenediamine, Carbomer, Fragrance
    Format Gel
    Shelf Life 24 Month(s)
    Product Type Stoko™ Instant Hand Sanitizer
    pH Range 6.8 to 7.9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit